Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VISTA/B7-H5/PD-1H Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$176.44 - $449.38

Specifications

Antigen VISTA/B7-H5/PD-1H
Clone 1011478
Dilution Blockade of Receptor-ligand Interaction
Applications Blocking Assay
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB71266SP
GREENER_CHOICE
View Documents
R&D Systems
MAB71266SP
25 μg
Each for $176.44
Only null left
Add to Cart
 
MAB71266100
GREENER_CHOICE
View Documents
R&D Systems
MAB71266100
100 μg
Each for $449.38
Only null left
Add to Cart
 
Description

Description

VISTA/B7-H5/PD-1H Monoclonal specifically detects VISTA/B7-H5/PD-1H in Human samples. It is validated for Blockade of Receptor-ligand Interaction.
Specifications

Specifications

VISTA/B7-H5/PD-1H
Blockade of Receptor-ligand Interaction
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Mouse myeloma cell line NS0-derived recombinant human VISTA/B7-H5/PD-1H, Phe33-Ala194, Accession # Q9H7M9
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human VISTA/B7-H5/PD-1H in direct ELISAs.
1011478
Blocking Assay
Unconjugated
Mouse
Human
Q9H7M9
64115
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.