Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Human VISTA/B7-H5/PD-1H DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human VISTA/B7-H5/PD-1H.
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Binding Target | VISTA/B7-H5/PD-1H |
| Assay Range | 23.40 to 1500 pg/mL |
| Format | 96-well |
| Gene ID (Entrez) | 64115 (Human); 74048 (Mouse); 690899 (Rat); 102116468 (Cynomolgus Monkey) |
| Sample Type | Cell Culture Supernatants, Serum, EDTA Plasma, Heparin Plasma |
| Synonym | 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1 H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
