Learn More
Human VMAT2 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB8327V100UG
Catalog No. FAB8327V100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
VMAT2 Monoclonal specifically detects VMAT2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
VMAT2
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q05940
SLC18A2
NS0 mouse myeloma cell line transfected with human VMAT2 Met1-Asp514 Accession # Q05940
RUO
6571
Human
IgG1
Flow Cytometry
899327
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
MGC120477, MNAT, monoamine neurotransmitter transporter, Monoamine transporter, SLC18A2, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2
Mouse
100 μg
Primary
Detects human VMAT2 in ELISAs and Western blots.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only (RUO)