Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VMAT2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8327
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8327 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8327 Supplier R&D Systems Supplier No. MAB8327
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VMAT2 Monoclonal specifically detects VMAT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen VMAT2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 899327
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q05940
Gene Alias MGC120477, MNAT, monoamine neurotransmitter transporter, Monoamine transporter, SLC18A2, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2
Gene Symbols SLC18A2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human VMAT2 Met1-Asp514 Accession # Q05940
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6571
Test Specificity Detects human VMAT2 in ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.