Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VPS26A Partial ORF (NP_004887.2, 228 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7098123
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: ETIAKYEIMDGAPVKGESIPIRLFLAGYDPTPTMRDVNKKFSVRYFLNLVLVDEEDRRYFKQQEIILWRKAPEKLRKQRTNFHQRFESPESQASAEQPEM

Specifications

Accession Number NP_004887.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9559
Molecular Weight (g/mol) 36.74kDa
Name VPS26A (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen ETIAKYEIMDGAPVKGESIPIRLFLAGYDPTPTMRDVNKKFSVRYFLNLVLVDEEDRRYFKQQEIILWRKAPEKLRKQRTNFHQRFESPESQASAEQPEM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ12930/HB58/Hbeta58/PEP8A/VPS26
Common Name VPS26A
Gene Symbol VPS26A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.