Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human VRK3 Partial ORF (NP_001020949, 322 a.a. - 424 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7100413 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-967-178 25 μg
89-967-177 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-967-178 Supplier Abnova™ Supplier No. H00051231Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. In both human and mouse, this gene has substitutions at several residues within the ATP binding motifs that in other kinases have been shown to be required for catalysis. In vitro assays indicate the protein lacks phosphorylation activity. The protein, however, likely retains its substrate binding capability. This gene is widely expressed in human tissues and its protein localizes to the nucleus. Alternative splicing results in multiple transcripts encoding different isoforms. [provided by RefSeq]

Sequence: DLQSLGYCMLKWLYGFLPWTNCLPNTEDIMKQKQKFVDKPGPFVGPCGHWIRPSETLQKYLKVVMALTYEEKPPYAMLRNNLEALLQDLRVSPYDPIGLPMVP

Specifications

Accession Number NP_001020949
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51231
Molecular Weight (g/mol) 37.07kDa
Name VRK3 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen DLQSLGYCMLKWLYGFLPWTNCLPNTEDIMKQKQKFVDKPGPFVGPCGHWIRPSETLQKYLKVVMALTYEEKPPYAMLRNNLEALLQDLRVSPYDPIGLPMVP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name VRK3
Gene Symbol VRK3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.