Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VSIG2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB51771SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB51771SP 25 μg
MAB51771100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB51771SP Supplier R&D Systems Supplier No. MAB51771SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VSIG2 Monoclonal specifically detects VSIG2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen VSIG2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 541522
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q961Q7-1
Gene Alias 2210413P10Rik, Cortical thymocyte-like protein, CTH, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, CTXL, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human VSIG2, Val24-Ala243, Accession # Q961Q7-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 23584
Test Specificity Detects human VSIG2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.