Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VSIG3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB92293SP
This item is not returnable.
View return policy
Description
VSIG3/IGSF11 Monoclonal antibody specifically detects VSIG3/IGSF11 in Human samples. It is validated for Flow Cytometry, Blocking Assay, CyTOFSpecifications
VSIG3/IGSF11 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Brain and testis-specific immunoglobulin superfamily protein, Bt-IGSF, BTIGSF, cancer/testis antigen 119, CT119, CXADR like 1, CXADRL1, IgSF11, Igsf13, immunoglobulin superfamily member 11, immunoglobulin superfamily, member 11, MGC35227, V-set and immunoglobulin domain containing 3, V-set and immunoglobulin domain-containing protein 3, VSIG3brain and testis-specific immunoglobin superfamily protein | |
Mouse myeloma cell line NS0-derived recombinant human VSIG3, Met1-Gly245, Accession # Q5DX21 | |
25 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, Blocking Assay, CyTOF | |
973422 | |
Flow Cytometry 0.25 ug/106 cells, Blockade of Receptor-ligand Interaction 0.2-2 ug/mL, CyTOF-ready | |
Q5DX21 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
152404 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction