Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VSIG4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $491.00
Specifications
| Antigen | VSIG4 |
|---|---|
| Clone | 2174B |
| Dilution | Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/106 cells |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB46461SP
![]() |
R&D Systems
MAB46461SP |
25 μg |
Each for $200.50
|
|
|||||
MAB46461100
![]() |
R&D Systems
MAB46461100 |
100 μg |
Each for $491.00
|
|
|||||
Description
VSIG4 Monoclonal antibody specifically detects VSIG4 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| VSIG4 | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/106 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily | |
| Mouse myeloma cell line NS0-derived human VSIG4, Arg20-Pro283, Accession # Q9Y279 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2174B | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9Y279 | |
| 11326 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title