Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Human VSIG4 ELISA Kit (Colorimetric)
SDP

Catalog No. NB228810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB228810 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB228810 Supplier Novus Biologicals Supplier No. NBP280355

Specifications

Assay Range 23.44-1500 pg/mL
Assay Sensitivity 12.63 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Lysates
Specificity Recognizes both recombinant and natural Human VSIG4.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 11326
Gene Symbol VSIG4
Quantity 1 Kit
Storage Requirements Storage is content dependent.
Synonym CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.