Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VSIG8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9418SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9418SP 25 μg
MAB9418100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9418SP Supplier R&D Systems Supplier No. MAB9418SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VSIG8 Monoclonal specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen VSIG8
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 961823
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q5VU13
Gene Alias V-set and immunoglobulin domain containing 8
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391123
Test Specificity Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.