Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VSIG8 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9418P100
This item is not returnable.
View return policy
Description
VSIG8 Monoclonal specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry.Specifications
VSIG8 | |
Monoclonal | |
PE | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
V-set and immunoglobulin domain containing 8 | |
Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13 | |
100 Tests | |
391123 | |
Human | |
Supernatant |
Flow Cytometry | |
961823 | |
Flow Cytometry 10μL/106 cells | |
Q5VU13 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction