Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VSTM2A Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB100371G1
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB100371G1 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB100371G1 Supplier R&D Systems Supplier No. FAB100371G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VSTM2A Monoclonal antibody specifically detects VSTM2A in Human samples. It is validated for Flow Cytometry

Specifications

Antigen VSTM2A
Applications Flow Cytometry
Classification Monoclonal
Clone 1024735
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8TAG5-2
Gene Alias MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2A, V-set and transmembrane domain-containing protein 2A, VSTM2
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human VSTM2A protein, Ser25-Phe244, Accession # Q8TAG5-2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222008
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.