Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human VSTM2A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB100371SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB100371SP 25 μg
MAB10037110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB100371SP Supplier R&D Systems Supplier No. MAB100371SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VSTM2A Monoclonal specifically detects VSTM2A in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen VSTM2A
Applications Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 1024735
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, ELISA, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. Q8TAG5-2
Gene Alias MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2A, V-set and transmembrane domain-containing protein 2A, VSTM2
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived human VSTM2A protein, Ser25-Phe244, Accession # Q8TAG5-2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222008
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.