Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VSTM4 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB10767R100UG
This item is not returnable.
View return policy
Description
VSTM4 Monoclonal antibody specifically detects VSTM4 in Human samples. It is validated for Flow CytometrySpecifications
VSTM4 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C10orf72, FLJ31737, hypothetical protein LOC196740, V-set and transmembrane domain containing 4 | |
Chinese Hamster Ovary cell line CHO-derived human VSTM4, Leu24-Tyr180, Accession # Q8IW00-1 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Flow Cytometry | |
2694A | |
Flow Cytometry 0.25-1 ug/106 cells | |
Q8IW00-1 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
196740 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction