Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human WASP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3070
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3070 100 μg
MAB3070SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3070 Supplier R&D Systems Supplier No. MAB3070
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

WASP Monoclonal specifically detects WASP in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen WASP
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 336925
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P42768
Gene Alias IMD2THC, Thrombocytopenia 1, WAS, WASP, WiskTHC1, wiskthrombocytopenia 1 (X-linked)
Gene Symbols WAS
Host Species Mouse
Immunogen E. coli-derived recombinant human WASP Lys230-Gln310 Accession # P42768
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7454
Test Specificity Detects human WASP in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.