Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human WIF-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF134SP
This item is not returnable.
View return policy
Description
WIF-1 Polyclonal specifically detects WIF-1 in Human samples. It is validated for Western Blot.Specifications
WIF-1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
WIF1, WIF-1, WNT inhibitory factor 1 | |
Goat | |
Affinity Purified | |
RUO | |
11197 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
AAD25402 | |
WIF1 | |
Mouse myeloma cell line NS0-derived recombinant human WIF-1 Gly29-Trp379 Accession # AAD25402 | |
25 μg | |
Primary | |
Detects human and mouse WIF-1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction