Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human WIF-1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 3 publications

$200.50 - $2350.00

Specifications

Antigen WIF-1
Clone 133015
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Applications Western Blot, Immunohistochemistry
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB134SP
GREENER_CHOICE
View Documents
R&D Systems
MAB134SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB134050
GREENER_CHOICE
View Documents
R&D Systems
MAB134050
50 μg
Each for $292.00
Only null left
Add to Cart
 
MAB134500
GREENER_CHOICE
View Documents
R&D Systems
MAB134500
500 μg
Each for $2,350.00
Only null left
Add to Cart
 
Description

Description

WIF-1 Monoclonal specifically detects WIF-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

WIF-1
LYOPH
Western Blot, Immunohistochemistry
Unconjugated
Mouse
Human
AAD25402
11197
Mouse myeloma cell line NS0-derived recombinant human WIF-1 Gly29-Trp379 Accession # AAD25402
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human WIF-1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse WIF-1 is observed.
133015
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
WIF1, WIF-1, WNT inhibitory factor 1
WIF1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.