Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human WISP-1/CCN4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1627
This item is not returnable.
View return policy
Description
WISP-1/CCN4 Monoclonal specifically detects WISP-1/CCN4 in Human samples. It is validated for Western Blot.Specifications
WISP-1/CCN4 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O95388 | |
WISP1 | |
Mouse myeloma cell line NS0-derived recombinant human WISP-1/CCN4 Thr23-Asn367 Accession # O95388 | |
500 μg | |
Primary | |
Detects human WISP-1/CCN4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) CTGF, rhNOV, or rmWISP-1. | |
Human | |
Purified |
Western Blot | |
213611 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCN family member 4, CCN4, CCN4WISP1tc, WISP1, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8840 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction