Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human WISP-1/CCN4 ELISA Kit (Colorimetric)

Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Assay Range | 78.13 to 5000 pg/mL |
Assay Sensitivity | 46.88 pg/mL |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum |
Specificity | This kit recognizes Human WISP-1/CCN4 in samples. No significant cross-reactivity or interference between Human WISP-1/CCN4 and analogues was observed. |
Target Species | Human |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 8840 |
Gene Symbol | CCN4 |
Quantity | 1 Kit |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction