Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human WNK4 Partial ORF (NP_115763.2, 1144 a.a. - 1243 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89958233
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the WNK family of serine-threonine protein kinases. The kinase is part of the tight junction complex in kidney cells, and regulates the balance between NaCl reabsorption and K(+) secretion. The kinase regulates the activities of several types of ion channels, cotransporters, and exchangers involved in electrolyte flux in epithelial cells. Mutations in this gene result in pseudohypoaldosteronism type IIB

Sequence: KHLSEVETLQTLQKKEIEDLYSRLGKQPPPGIVAPAAMLSSRQRRLSKGSFPTSRRNSLQRSEPPGPGIMRRNSLSGSSTGSQEQRASKGVTFAGDVGRM

Specifications

Accession Number NP_115763.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 65266
Molecular Weight (g/mol) 36.74kDa
Name WNK4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen KHLSEVETLQTLQKKEIEDLYSRLGKQPPPGIVAPAAMLSSRQRRLSKGSFPTSRRNSLQRSEPPGPGIMRRNSLSGSSTGSQEQRASKGVTFAGDVGRM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias PHA2B/PRKWNK4
Common Name WNK4
Gene Symbol WNK4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.