Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Wnt-4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6076
This item is not returnable.
View return policy
Description
Wnt-4 Polyclonal specifically detects Wnt-4 in Human samples. It is validated for Western Blot.Specifications
Wnt-4 | |
Polyclonal | |
Unconjugated | |
P56705 | |
WNT4 | |
Chinese hamster ovary cell line CHO-derived recombinant human Wnt-4 Ser23-Arg351 Accession # P56705 | |
100 μg | |
Primary | |
Detects human Wnt-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 10-15% cross-reactivity with recombinant human Wnt-3A and recombinant mouse Wnt-10B is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
protein Wnt-4, SERKAL, wingless-type MMTV integration site family, member 4, Wnt4, WNT-4 | |
Sheep | |
Affinity Purified | |
RUO | |
54361 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction