Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Wnt-7a/b Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF3008
This item is not returnable.
View return policy
Description
Wnt-7a/b Polyclonal specifically detects Wnt-7a/b in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt-7a/b | |
Polyclonal | |
Unconjugated | |
O00755 | |
Goat | |
Affinity Purified | |
RUO | |
7476 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Wnt7a, Wnt-7a, Wnt7b | |
E. coli-derived recombinant human Wnt-7a Leu32-Glu72, Thr216-Arg292 Accession # O00755 | |
100 μg | |
Primary | |
Detects human Wnt-7a and Wnt-7b in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Wnt-1, rmWnt-3a, rmWnt-4, rmWnt-5a, rmWnt-5b, rmWnt-9a, rmWnt-11, and rmWnt-16 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction