Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Wnt-7b Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3460
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3460 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3460 Supplier R&D Systems Supplier No. AF3460
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 7 publications

Wnt-7b Polyclonal specifically detects Wnt-7b in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Wnt-7b
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P56706
Gene Alias protein Wnt-7b, wingless-type MMTV integration site family, member 7B, Wnt7b
Gene Symbols WNT7B
Host Species Goat
Immunogen E. coli-derived recombinant human Wnt-7b Leu32-Glu72 & Thr216-Ala283 Accession # P56706
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7477
Test Specificity Detects human Wnt-7b in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Wnt-7a is observed and less than 5% cross-reactivity with recombinant mouse (rm) Wnt-1, rmWnt-3a, rmWnt-4, rmWnt-5a, rhWnt-9a, rmWnt-11, and rmWnt-16 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.