Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Wnt-9b Recombinant Protein, PeproTech®

Catalog No. 12049100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
20 μg
5 μg
5 x 100 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12049100UG 100 μg
12-049-500 5 x 100 μg
120-49-20UG 20 μg
120-49-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 12049100UG Supplier Gibco™ Supplier No. 12049100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

120-49-1MG will be provided as 10 x 100 μg (120-49-100UG). 120-49-250UG will be provided as multiple 50 μg (120-49-50UG) and 100 μg (120-49-100UG) vials. 120-49-500UG will be provided as 5 x 100 μg (120-49-100UG). Recombinant Human Wnt-9b is a monomeric glycoprotein containing 335 amino acid residues and has a calculated molecular weight of 36.9 kDa. Due to glycosylation, Recombinant Human Wnt-9b migrates at an apparent molecular weight of approximately 49-54 kDa by SDS-PAGE analysis under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

WNT9B, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14905
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 7484
Molecular Weight (g/mol) 36.9 kDa
Name Human Wnt-9 b
Quantity 100 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias clf; clf1; protein Wnt-14 b; Protein Wnt-15; Protein Wnt-9 b; UNQ6973/PRO21956; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9 B; wingless-type MMTV integration site family member 9 B; wingless-type MMTV integration site family, member 15; wingless-type MMTV integration site family, member 9 B; Wnt family member 9 B; Wnt14b; wnt-14 b; Wnt15; wnt-15; Wnt9b
Common Name WNT9B
Gene Symbol Wnt9b
Biological Activity Determined by its ability to induce alkaline phosphatase production by CCL-226 cells.
Conjugate Unconjugated
Recombinant Recombinant
Sequence SYFGLTGREV LTPFPGLGTA AAPAQGGAHL KQCDLLKLSR RQKQLCRREP GLAETLRDAA HLGLLECQFQ FRHERWNCSL EGRMGLLKRG FKETAFLYAV SSAALTHTLA RACSAGRMER CTCDDSPGLE SRQAWQWGVC GDNLKYSTKF LSNFLGSKRG NKDLRARADA HNTHVGIKAV KSGLRTTCKC HGVSGSCAVR TCWKQLSPFR ETGQVLKLRY DSAVKVSSAT NEALGRLELW APARQGSLTK GLAPRSGDLV YMEDSPSFCR PSKYSPGTAG RVCSREASCS SLCCGRGYDT QSRLVAFSCH CQVQWCCYVE CQQCVQEELV YTCKH
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.