Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human XBP1 (S Isoform) Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC4257N100UG
This item is not returnable.
View return policy
Description
XBP1 Monoclonal specifically detects XBP1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
XBP1 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_001073007 | |
XBP1 | |
E. coli-derived recombinant human XBP1S Ser188-Leu287 Accession # NP_001073007 | |
RUO | |
7494 | |
Human | |
IgG1 |
Flow Cytometry | |
525904 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
HTF, TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2, XBP2Tax-responsive element-binding protein 5 | |
Mouse | |
100 μg | |
Primary | |
Detects human XBP1 (S Isoform) (XBP1S) in direct ELISAs. This segment is found only in the XBP1S (spliced) variant. Does not detect XBP1U (unspliced). | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction