Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Xg Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3699
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3699 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3699 Supplier R&D Systems Supplier No. AF3699
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Xg Polyclonal specifically detects Xg in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen Xg
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells
Gene Accession No. P55808
Gene Alias glycoprotein Xg, MGC118758, MGC118759, MGC118760, MGC118761, PBDX, Protein PBDX, Xg blood group, Xg blood group (pseudoautosomal boundary-divided on the X chromosome), XG glycoprotein, XGPY
Gene Symbols XG
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Xg Gln22-Lys142 Accession # P55808
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7499
Test Specificity Detects human Xg in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.