Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human XPA Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3416
This item is not returnable.
View return policy
Description
XPA Polyclonal specifically detects XPA in Human samples. It is validated for Western Blot.Specifications
| XPA | |
| Polyclonal | |
| Unconjugated | |
| P23025 | |
| XPA | |
| E. coli-derived recombinant human XPA Met1-Met273 Accession # P23025 | |
| 100 μg | |
| Primary | |
| Detects human XPA in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| DNA repair protein complementing XP-A cells, excision repair-controlling, Xeroderma pigmentosum group A-complementing protein, xeroderma pigmentosum, complementation group A, XP1, XPACXP1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7507 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction