Learn More
Human ZAP70 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | ZAP70 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Gene Accession No. | P43403 |
| Gene Alias | 70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, mrtle, mur, SRK, SRKFLJ17679, STD, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD |
| Gene Symbols | ZAP70 |
| Host Species | Sheep |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.