Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ZBTB38 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5784SP
This item is not returnable.
View return policy
Description
ZBTB38 Polyclonal specifically detects ZBTB38 in Human samples. It is validated for Western Blot.Specifications
ZBTB38 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CIBZ, FLJ22332, FLJ31131, FLJ35036, STQTL10, zinc finger and BTB domain containing 38, zinc finger and BTB domain-containing protein 38, ZNF921 | |
Goat | |
Affinity Purified | |
RUO | |
253461 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q8NAP3 | |
ZBTB38 | |
E. coli-derived recombinant human ZBTB38 Met1-Tyr232 Accession # Q8NAP3 | |
25 μg | |
Primary | |
Detects endogenous human ZBTB38 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction