Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ZEB2/SIP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $503.00
Specifications
Antigen | ZEB2 |
---|---|
Clone | 923328 |
Dilution | Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Immunohistochemistry, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB73782SP
![]() |
R&D Systems
MAB73782SP |
25 μg |
Each for $214.00
|
|
|||||
MAB73782
![]() |
R&D Systems
MAB73782 |
100 μg |
Each for $503.00
|
|
|||||
Description
ZEB2 Monoclonal specifically detects ZEB2 in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
ZEB2 | |
Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HSPC082, KIAA0569FLJ42816, SIP1, SIP1HSPC082, Smad-interacting protein 1, SMADIP1, SMADIP1ZFHX1BSIP-1, ZFHX1B, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b | |
ZEB2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
923328 | |
Immunohistochemistry, Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
O60315 | |
9839 | |
E. coli-derived recombinant human ZEB2/SIP1 Asn363-Lys537 Accession # O60315 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ZEB2/SIP1 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title