Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ZFP90 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7870SP
This item is not returnable.
View return policy
Description
ZFP90 Polyclonal specifically detects ZFP90 in Human samples. It is validated for Western Blot.Specifications
ZFP90 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FoxP3-interacting Protein, KIAA1954, NK10, zfp-90, zinc finger protein 476, zinc finger protein 756, zinc finger protein 90 homolog, zinc finger protein 90 homolog (mouse), ZNF756 | |
Sheep | |
Affinity Purified | |
RUO | |
146198 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q8TF47 | |
ZFP90 | |
E. coli-derived recombinant human ZFP90 Lys94-Lys207 Accession # Q8TF47 | |
25 μg | |
Primary | |
Detects human ZFP90 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse ZFP90 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction