Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ZFP90 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7870
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7870 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7870 Supplier R&D Systems Supplier No. AF7870
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ZFP90 Polyclonal specifically detects ZFP90 in Human samples. It is validated for Western Blot.

Specifications

Antigen ZFP90
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q8TF47
Gene Alias FoxP3-interacting Protein, KIAA1954, NK10, zfp-90, zinc finger protein 476, zinc finger protein 756, zinc finger protein 90 homolog, zinc finger protein 90 homolog (mouse), ZNF756
Gene Symbols ZFP90
Host Species Sheep
Immunogen E. coli-derived recombinant human ZFP90 Lys94-Lys207 Accession # Q8TF47
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146198
Test Specificity Detects human ZFP90 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse ZFP90 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.