Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ZNF10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB96561100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB96561100 100 μg
MAB96561SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB96561100 Supplier R&D Systems Supplier No. MAB96561100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF10 Monoclonal specifically detects ZNF10 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen ZNF10
Applications Immunohistochemistry
Classification Monoclonal
Clone 980803
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P21506
Gene Alias KOX1, KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1
Host Species Mouse
Immunogen Synthetic peptide containing human ZNF10 Accession # P21506
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7556
Test Specificity Detects human ZNF10 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.