Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ZNF10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | ZNF10 |
---|---|
Clone | 980809 |
Dilution | Western Blot 0.5 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9656SP
![]() |
R&D Systems
MAB9656SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9656100
![]() |
R&D Systems
MAB9656100 |
100 μg |
Each for $502.50
|
|
|||||
Description
ZNF10 Monoclonal specifically detects ZNF10 in Human samples. It is validated for Western Blot.Specifications
ZNF10 | |
Western Blot 0.5 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
P21506 | |
7556 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
980809 | |
Western Blot | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
KOX1, KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1 | |
Synthetic peptide containing human ZNF10 Accession # P21506 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ZNF10 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title