Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ZNF206 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4020
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4020 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4020 Supplier R&D Systems Supplier No. AF4020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

ZNF206 Polyclonal specifically detects ZNF206 in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen ZNF206
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q96SZ4
Gene Alias Zfp206, ZSCAN10, ZSCAN10 zinc finger and SCAN domain containing 10
Gene Symbols ZSCAN10
Host Species Goat
Immunogen E. coli-derived recombinant human ZNF206 Met1-Val283 Accession # Q96SZ4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84891
Test Specificity Detects human ZNF206 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human ZNF24 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.