Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ZNF331 Partial ORF (NP_061025.4, 362 a.a. - 459 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. p-7101043
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form one family of zinc finger proteins. See ZFP93 (MIM 604749) for additional information on zinc finger proteins.[supplied by OMIM]

Sequence: KAFNCGYHLTQHERIHTGETPYKCKECGKAFIYGSSLVKHERIHTGVKPYGCTECGKSFSHGHQLTQHQKTHSGAKSYECKECGKACNHLNHLREHQR

Specifications

Accession Number NP_061025.4
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55422
Molecular Weight (g/mol) 36.52kDa
Name ZNF331 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen KAFNCGYHLTQHERIHTGETPYKCKECGKAFIYGSSLVKHERIHTGVKPYGCTECGKSFSHGHQLTQHQKTHSGAKSYECKECGKACNHLNHLREHQR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp686L0787/RITA/ZNF361/ZNF463
Common Name ZNF331
Gene Symbol ZNF331
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.