Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ZnT-8 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7318
This item is not returnable.
View return policy
Description
ZnT-8/SLC30A8 Polyclonal specifically detects ZnT-8/SLC30A8 in Human samples. It is validated for Immunohistochemistry.Specifications
ZnT-8/SLC30A8 | |
Polyclonal | |
Unconjugated | |
Q8IWU4 | |
SLC30A8 | |
E. coli-derived recombinant human ZnT-8 Lys268-Pro359 Accession # Q8IWU4 | |
100 μg | |
Primary | |
Detects human Znt-8 in direct ELISAs. | |
Human | |
IgG |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
SLC30A8, solute carrier family 30 (zinc transporter), member 8, Solute carrier family 30 member 8, zinc transporter 8, zinc transporter ZnT-8, ZnT8, ZNT8ZnT-8 | |
Sheep | |
Affinity Purified | |
RUO | |
169026 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction