Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Humanin Antibody, Novus Biologicals™
SDP

Catalog No. p-5074155 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056876T 0.025 mL
NB10056876 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056876T Supplier Novus Biologicals Supplier No. NB10056876SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Humanin Polyclonal specifically detects Humanin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Humanin
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000, ELISA 1:100-1:1000
Formulation Whole antisera with 50% Glycerol with 0.025% Sodium Azide
Gene Accession No. Q8IVG9
Gene Alias HN, HUNIN
Gene Symbols MTRNR2L1
Host Species Rabbit
Immunogen Amino acids 12-24 of human humanin protein were used as the immunogen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100462977
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.