Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Humanin Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300245PE
Description
Humanin Polyclonal antibody specifically detects Humanin in Human samples. It is validated for Western BlotSpecifications
Humanin | |
Polyclonal | |
Western Blot | |
HN, HUNIN | |
Residues 9-24 (LLLLTSEIDLPVKRRA) of the human Humanin protein. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
100462977 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction