Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Humic acid

Catalog No. 5368010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
50 g

Catalog No. 53-680-10G

Supplier: Sigma Aldrich 5368010G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Humic acid (ha) is a heterogeneous macromolecule mainly found in soil and water.humic acid has been used to prepare stock solution employed in the following experiments:
  • Coagulation activity assay of moringa oleifera protein extracts for humic acid removal
  • Adsorption edge experiment to study its effect on the sorption of tetracycline onto goethite
  • Evaluation of the ability of polymeric adsorbent supelite(tm) dax-8 to remove humic acid from nucleic acid extracts prior to quantitative real-time pcr (qpcr).supelite is a trademark of sigma-aldrich co. llc

Specifications

CAS 1415-93-6
MDL Number MFCD00147177
Synonym PGS 10
Quantity 10 g
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.