Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Humic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
50 g
Description
- Humic acid (ha) is a heterogeneous macromolecule mainly found in soil and water.humic acid has been used to prepare stock solution employed in the following experiments:
- Coagulation activity assay of moringa oleifera protein extracts for humic acid removal
- Adsorption edge experiment to study its effect on the sorption of tetracycline onto goethite
- Evaluation of the ability of polymeric adsorbent supelite(tm) dax-8 to remove humic acid from nucleic acid extracts prior to quantitative real-time pcr (qpcr).supelite is a trademark of sigma-aldrich co. llc
Specifications
Specifications
| CAS | 1415-93-6 |
| MDL Number | MFCD00147177 |
| Synonym | PGS 10 |
| Quantity | 50 g |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction