Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Humic acid, sodium salt, 45-70%, technical, (as humic acid)

Catalog No. ac120860050
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle

CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68131-04-4
Molecular Formula C9H8Na2O4
Molecular Weight (g/mol) 226.139
MDL Number MFCD00135560
InChI Key HMGUIQPKFUZDPV-UHFFFAOYSA-L
Synonym humic acid sodium salt, sodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, sodium humate, disodium bicyclo 2.2.1 hept-5-en-2,3-dicarboxylate, disodium bicyclo 2.2.1 hept-5-ene-2,3-dicarboxylate, bicyclo 2.2.1 hepta-5-ene-2,3-dicarboxylic acid disodium salt
PubChem CID 21908873
IUPAC Name disodium;bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
SMILES C1C2C=CC1C(C2C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Color Black
Assay Percent Range 45-70%
Percent Purity 45 to 70%
Grade Technical
Quantity 5 g
Infrared Spectrum Authentic
Loss on Drying 14% to 25% (105°C, 4 hrs)
Packaging Glass bottle
Physical Form Crystalline Powder
Chemical Name or Material Humic acid, sodium salt, tech., 50-60% (as humic acid)
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 268-608-0
RTECSNumber MT6550000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.