Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Humicade Chimeric Recombinant Rabbit Monoclonal Antibody (CDP 571 (Humicade))
GREENER_CHOICE

Catalog No. PIMA547967
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547967 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547967 Supplier Invitrogen™ Supplier No. MA547967
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: The antibody binds to human TNF-alpha with a Kd of 0.087 nM.

The structure of Humicade is a monoclonal antibody that is highly similar to the reference medicine, Humira (adalimumab). It is composed of human immunoglobulin G1 (IgG1) antibodies that are produced using recombinant DNA technology. The function of Humicade is to treat certain autoimmune conditions by blocking the action of tumor necrosis factor-alpha (TNF-alpha), a protein that contributes to inflammation. The structure of Humicade allows it to specifically target TNF-alpha and block its activity, reducing inflammation and alleviating symptoms in certain autoimmune conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Humicade Chimeric
Applications ELISA, Neutralization
Classification Recombinant Monoclonal
Clone CDP 571 (Humicade)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Host Species Rabbit
Immunogen Human TNF-alpha.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.