Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HUNK, GST Tag Recombinant Protein

Catalog No. A32892
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg

Recombinant Protein

HUNK serine/threonine kinase or hormonally up-regulated Neu-associated kinase is a member of the mitogen-activated protein kinase family which is essential for mammary tumor metastasis. HUNK deletion in mice did not alter viability, fertility, longevity, or propensity to develop tumors and it is also dispensable for murine development. HUNK has been shown to participate in vesicular transport. Additionally, functional studies in mice suggest that HUNK regulates endocytosis via interaction with rabaptin-5 protein and induces changes in mammary glands during pregnancy. HUNK expression has been documented in mouse brain, breast, embryo, and fetus. ESTs have been isolated from several human tissue libraries, including normal ear.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P57058
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 30811
Molecular Weight (g/mol) 105.9 kDa
Name Human HUNK, GST Tag
Quantity 100 μg
Gene Alias B19; Bstk1; Hormonally up-regulated neu tumor-associated kinase; hormonally upregulated neu tumor-associated kinase; hormonally up-regulated Neu-associated kinase; hormonally upregulated Neu-associated kinase; Hunk; MAKV; Mak-v; RGD1565091; serine/threonine protein kinase MAK-V; serine/threonine-protein kinase MAK-V
Common Name HUNK
KinaseFamily CAMKL Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.