Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Huntingtin Antibody (HDB4E10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001197
Description
Huntingtin Monoclonal specifically detects Huntingtin in Human, Mouse, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
Huntingtin | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
HD protein, huntingtin, huntingtin (Huntington disease), IT15HDHuntington disease protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This reacts with an epitope corresponding to the HDB region (amino acids 1844 - 2131) of the huntingtin protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
HDB4E10 | |
Unconjugated | |
P42858 | |
HTT | |
Recombinant fragment, corresponding to amino acids 1844-2131 of Huntingtin. | |
0.05 mg | |
Apoptosis, Autophagy, Cytoskeleton Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
3064 | |
Human, Mouse, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction