Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Huntingtin Interacting Protein K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $792.00
Specifications
Antigen | Huntingtin Interacting Protein K |
---|---|
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Huntingtin Interacting Protein K Polyclonal specifically detects Huntingtin Interacting Protein K in Human, Rat samples. It is validated for Western Blot.Specifications
Huntingtin Interacting Protein K | |
Western Blot | |
Unconjugated | |
RUO | |
No buffer with No Preservative | |
C15orf63, chromosome 15 open reading frame 63, EC 2.1.1.43, EC 2.7.7.6, FLJ20431, HSPC136, Huntingtin yeast partner K, huntingtin-interacting protein K, HYPKHuntingtin interacting protein K | |
HYPK | |
IgG | |
Affinity Purified | |
Huntingtin-interacting protein 1 (HIP1). By Western Blot the antibody recognizes a band at ∼116 kDa on P2 microsomal fraction from embryonic rat brain. |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Rat, Human | |
O00291 | |
25764 | |
Synthetic peptide. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
116 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title