Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HuR/ELAVL1 Antibody, Janelia Fluor™ 525, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298790JF525
Description
HuR/ELAVL1 Polyclonal antibody specifically detects HuR/ELAVL1 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
HuR/ELAVL1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
Janelia Fluor 525 | |
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R), ELAV-like protein 1, embryonic lethal, abnormal vision, drosophila, homolog-like 1, Hu antigen R, Hua, Hu-antigen R, HuR, HURELAV1, MelG | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human HuR/ELAVL1. | |
0.1 mL | |
Vision | |
1994 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction