Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HuR/ELAVL1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298790MFV610
Description
HuR/ELAVL1 Polyclonal antibody specifically detects HuR/ELAVL1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| HuR/ELAVL1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R), ELAV-like protein 1, embryonic lethal, abnormal vision, drosophila, homolog-like 1, Hu antigen R, Hua, Hu-antigen R, HuR, HURELAV1, MelG | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human HuR/ELAVL1. | |
| 0.1 mL | |
| Vision | |
| 1994 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction