Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HuR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590917
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590917 Supplier Invitrogen™ Supplier No. PA590917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSNGYEDHMA EDCRGDIGRT NLIVNYLPQN MTQDELRSLF SSIGEVESAK LIRDKVAGHS LGYGFVNYVT AKDAERAINT LNGLRLQSKT IKVSYARPSS EVIKDANLYI SGLPRTMTQK DVEDMFSRFG RIINSRVLVD QTTGLSRGVA FIRFDKRSEA EEAITSFNGH KPPGSSEPIT VKFAANPNQN KNVALLSQLY HSPARRFGGP VHHQAQRFRF SPMGVDHMSG LSGVNVPGNA SSGWCIFIYN LGQDADEGIL WQMFGPFGAV TNVKVIRDFN TNKCKGFGFV TMTNYEEAAM AIASLNGYRL GDKILQVSFK TNKSHK; Positive Samples: U-251MG, HeLa, Jurkat, K-562, PC-3; Cellular Location: Cytoplasm, Nucleus.

The protein encoded by this gene is a member of the ELAVL protein family. This encoded protein contains 3 RNA-binding domains and binds cis-acting AU-rich elements. It destabilizes mRNAs and thereby regulates gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HuR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ELAVL1
Gene Accession No. B5DF91, P70372, Q15717
Gene Alias 2410055N02Rik; ELAV (embryonic lethal, abnormal vision)-like 1 (Hu antigen R); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 1 (Hu antigen R); ELAV like 1; ELAV like RNA binding protein 1; ELAV like RNA binding protein 1a; ELAV1; Elavl1; elavl1 protein; elavl1a; elav-like generic protein; ELAV-like protein 1; ELAV-like protein 1; ELAV-like protein 1a; elav-related A; Elra; embryonic lethal, abnormal vision, drosophila, homolog-like 1; embryonic lethal, abnormal vision-related A; etID309757.2; fa25f08; fb59a02; fc60g02; Hu antigen R; Hua; HU-antigen A; hu-antigen R; Human antigen R; HuR; melG; ribonucleoprotein; W91709; wu:fa25f08; wu:fb59a02; wu:fc60g02; zHuA
Gene Symbols ELAVL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-326 of human HuR/ELAVL1 (NP_0014102).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15568, 1994, 363854
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.